Assay Details
Target Gene Details
| Entrez Gene ID: | 2287 | 
| Gene Name: | FK506 binding protein 3 | 
| Gene Aliases: | FKBP-25, FKBP-3, FKBP25, PPIase | 
| Location: | Chr.14:45115599-45134529 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 4 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| FKBP3 | NM_002013.3 | 4 | 463 | NP_002004.1 | 
| XM_011536565.2 | 4 | 2484 | XP_011534867.1 | |
| XM_017021092.1 | 4 | 412 | XP_016876581.1 | |
| XM_017021093.1 | 4 | 2484 | XP_016876582.1 | |
| AK222993.1 | 4 | 412 | BAD96713.1 | |
| AK311915.1 | 4 | 401 | ||
| BC016288.1 | 5 | 801 | AAH16288.1 | |
| BC020809.1 | 4 | 401 | AAH20809.1 | |
| BI598048.1 | 4 | 468 | ||
| BT006904.1 | 4 | 390 | AAP35550.1 | |
| M96256.1 | 4 | 413 | AAA58471.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv832785 | Chr.14:45051467 - 45241538 on Build GRCh38 | Loss |  SNORD127  MIS18BP1  PRPF39  FAM179B  LOC105370474  FANCM  FKBP3 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top