Assay Details
Target Gene Details
Entrez Gene ID: | 1371 |
Gene Name: | coproporphyrinogen oxidase |
Gene Aliases: |
CPO, CPX, HCP |
Location: |
Chr.3:98568448-98593684 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPOX | NM_000097.5 | 7 | 2594 | NP_000088.3 |
| XM_005247125.4 | XP_005247182.1 | |||
| BC017210.2 | 7 | 2545 | AAH17210.1 | |
| BC023551.2 | 7 | 2580 | AAH23551.1 | |
| BC023554.2 | 7 | 2541 | AAH23554.1 | |
| BX648974.1 | 2 | 1999 | ||
| D16611.1 | 7 | 2280 | BAA04033.1 | |
| Z28409.1 | 7 | 2187 | CAA82250.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834775 | Chr.3:98451215 - 98648708 on Build GRCh38 | Gain |
|
| nsv591065 | Chr.3:98485354 - 99047482 on Build GRCh38 | Gain |
|
| dgv293n21 | Chr.3:98382257 - 98598992 on Build GRCh38 | Loss |
|
| dgv4819n100 | Chr.3:98565439 - 98662629 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map