Assay Details
Target Gene Details
Entrez Gene ID: | 9587 |
Gene Name: | MAD2L1 binding protein |
Gene Aliases: |
CMT2 |
Location: |
Chr.6:43629542-43640952 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAD2L1BP | NM_001003690.1 | 4 | 699 | NP_001003690.1 |
| NM_014628.2 | 3 | 431 | NP_055443.1 | |
| AK297172.1 | 3 | 452 | ||
| AL537692.3 | 4 | 701 | ||
| BC002904.2 | 3 | 401 | AAH02904.1 | |
| BE297042.1 | ||||
| BI255998.1 | 3 | 418 | ||
| BU845738.1 | 4 | 736 | ||
| CB991410.1 | 3 | 458 | ||
| CR456893.1 | 3 | 395 | CAG33174.1 | |
| D14811.1 | 3 | 398 | BAA03552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5281 | Chr.6:43611090 - 43655905 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map