Assay Details
Target Gene Details
Entrez Gene ID: | 10058 |
Gene Name: | ATP binding cassette subfamily B member 6 (Langereis blood group) |
Gene Aliases: |
ABC, ABC14, DUH3, LAN, MCOPCB7, MTABC3, PRP, PSHK2, umat |
Location: |
Chr.2:219209766-219218990 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB6 | NM_005689.2 | NP_005680.1 | ||
| AF070598.1 | AAC28653.1 | |||
| AF076775.1 | AAF75107.1 | |||
| AF308472.1 | AAG33617.1 | |||
| AJ289233.2 | CAB95766.2 | |||
| AK026067.1 | ||||
| AK057026.1 | BAB71347.1 | |||
| AK172812.1 | BAD18782.1 | |||
| BC000559.2 | AAH00559.1 | |||
| BC043423.1 | AAH43423.1 | |||
| U66673.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893555 | Chr.2:219172722 - 219273269 on Build GRCh38 | Loss |
|
| nsv584500 | Chr.2:219210071 - 219224963 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map