Assay Details
Target Gene Details
Entrez Gene ID: | 9454 |
Gene Name: | homer scaffolding protein 3 |
Gene Aliases: |
HOMER-3, VESL3 |
Location: |
Chr.19:18929201-18941258 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOMER3 | NM_001145721.1 | NP_001139193.1 | ||
| NM_001145722.1 | NP_001139194.1 | |||
| NM_001145724.1 | NP_001139196.1 | |||
| NM_004838.3 | NP_004829.3 | |||
| NR_027297.1 | ||||
| XM_006722943.1 | XP_006723006.1 | |||
| XM_006722944.1 | XP_006723007.1 | |||
| AF093265.1 | AAC71029.1 | |||
| AK124450.1 | ||||
| AK313995.1 | ||||
| BC012113.1 | AAH12113.1 | |||
| BT019464.1 | AAV38271.1 | |||
| Y17573.1 | CAB75536.1 | |||
| Y18894.1 | CAB75543.1 | |||
| Y18895.1 | CAB75544.1 | |||
| Y18896.1 | CAB75545.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470132 | Chr.19:18853633 - 18943911 on Build GRCh38 | Loss |
|
| nsv833778 | Chr.19:18901860 - 19090678 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
| nsv952423 | Chr.19:18859292 - 18941091 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map