Assay Details
Target Gene Details
Entrez Gene ID: | 10309 |
Gene Name: | cyclin O |
Gene Aliases: |
CCNU, CILD29, UDG2 |
Location: |
Chr.5:55231152-55233717 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNO | NM_021147.4 | NP_066970.3 | ||
| NR_125346.1 | ||||
| NR_125347.1 | ||||
| NR_125348.1 | ||||
| AK026075.1 | BAB15351.1 | |||
| BC004877.2 | ||||
| X52486.1 | CAA36728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830302 | Chr.5:55107550 - 55259351 on Build GRCh38 | Gain |
|
| nsv1027090 | Chr.5:55179438 - 55375973 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76296175] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map