Assay Details
Target Gene Details
Entrez Gene ID: | 51654 |
Gene Name: | CDK5 regulatory subunit associated protein 1 |
Gene Aliases: |
C20orf34, C42, CGI-05, HSPC167 |
Location: |
Chr.20:33358839-33402768 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK5RAP1 | NM_001278167.1 | 14 | 2057 | NP_001265096.1 |
| NM_001278168.1 | 13 | 1829 | NP_001265097.1 | |
| NM_001278169.1 | 14 | 1828 | NP_001265098.1 | |
| NM_016082.4 | 14 | 1912 | NP_057166.4 | |
| NM_016408.3 | 14 | 2060 | NP_057492.2 | |
| XM_011528856.2 | 11 | 2796 | XP_011527158.1 | |
| XM_011528857.2 | 13 | 1672 | XP_011527159.1 | |
| XM_011528858.1 | 13 | 1911 | XP_011527160.1 | |
| XM_011528859.2 | 13 | 1671 | XP_011527161.1 | |
| XM_011528860.1 | 11 | 1371 | XP_011527162.1 | |
| XM_017027876.1 | 13 | 1810 | XP_016883365.1 | |
| XM_017027877.1 | 8 | 1124 | XP_016883366.1 | |
| AA721451.1 | 1 | 84 | ||
| AF152097.1 | 15 | 2038 | AAD34147.1 | |
| AF161516.1 | 14 | 2008 | AAF29131.1 | |
| AK023992.1 | 14 | 2010 | BAB14760.1 | |
| AK027449.1 | 14 | 1860 | BAB55120.1 | |
| AK095644.1 | 4 | 2587 | ||
| AK222745.1 | 14 | 2007 | BAD96465.1 | |
| AK292075.1 | 14 | 1778 | ||
| AK295342.1 | 14 | 1996 | ||
| BC001215.1 | 13 | 1758 | AAH01215.1 | |
| BC050706.1 | 14 | 2003 | AAH50706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064413 | Chr.20:33348381 - 33599978 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map