Assay Details
Target Gene Details
Entrez Gene ID: | 51755 |
Gene Name: | cyclin dependent kinase 12 |
Gene Aliases: |
CRK7, CRKR, CRKRS |
Location: |
Chr.17:39461486-39567560 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827985 | Chr.17:39461255 - 39463008 on Build GRCh38 | Loss |
|
| nsv1063848 | Chr.17:39419369 - 39551339 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map