Assay Details
Target Gene Details
Entrez Gene ID: | 8774 |
Gene Name: | NSF attachment protein gamma |
Gene Aliases: |
GAMMASNAP |
Location: |
Chr.18:10525876-10552769 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAPG | NM_003826.2 | 12 | 1172 | NP_003817.1 |
| XM_011525754.2 | 13 | 1256 | XP_011524056.1 | |
| XM_011525756.2 | 10 | 811 | XP_011524058.1 | |
| XM_017026063.1 | 8 | 728 | XP_016881552.1 | |
| AK126942.1 | 7 | 1325 | ||
| AK294038.1 | 11 | 951 | ||
| AK314495.1 | ||||
| BC001889.1 | 12 | 1008 | AAH01889.1 | |
| CR536554.1 | CAG38791.1 | |||
| U78107.1 | 12 | 1018 | AAB69753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059110 | Chr.18:10142986 - 10596015 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map