Assay Details
Target Gene Details
Entrez Gene ID: | 57567 |
Gene Name: | zinc finger protein 319 |
Gene Aliases: |
ZFP319 |
Location: |
Chr.16:57994668-58000691 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF319 | NM_020807.2 | 2 | 1258 | NP_065858.1 |
| XM_005256069.3 | 2 | 1183 | XP_005256126.1 | |
| AB037809.1 | 2 | 1258 | ||
| BC093919.1 | 1 | 697 | AAH93919.1 | |
| BC101767.1 | 1 | 697 | AAI01768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063386 | Chr.16:57958821 - 58040306 on Build GRCh38 | Gain |
|
| nsv1816 | Chr.16:57973452 - 58018171 on Build GRCh38 | Deletion |
|
| nsv572759 | Chr.16:57972385 - 58040306 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map