Assay Details
Target Gene Details
Entrez Gene ID: | 3182 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A/B |
Gene Aliases: |
ABBP1, HNRPAB |
Location: |
Chr.5:178204507-178211183 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPAB | NM_004499.3 | NP_004490.2 | ||
| NM_031266.2 | NP_112556.2 | |||
| AK054600.1 | ||||
| AK097657.1 | BAC05134.1 | |||
| AK223425.1 | BAD97145.1 | |||
| BC001616.1 | AAH01616.1 | |||
| BC002625.2 | AAH02625.1 | |||
| BC004561.2 | AAH04561.1 | |||
| BC009359.2 | AAH09359.1 | |||
| BC036708.2 | AAH36708.1 | |||
| M65028.1 | AAA36575.1 | |||
| U76713.1 | AAC50956.1 | |||
| Z36844.1 |
Target Gene Details
Entrez Gene ID: | 85007 |
Gene Name: | 5-phosphohydroxy-L-lysine phospho-lyase |
Gene Aliases: |
AGXT2L2, PHLU |
Location: |
Chr.5:178208474-178232822 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHYKPL | NM_001278346.1 | 13 | 2561 | NP_001265275.1 |
| NM_153373.3 | 13 | 1738 | NP_699204.1 | |
| NR_103508.1 | 10 | 1324 | ||
| NR_103509.1 | 10 | 2156 | ||
| NR_103510.1 | 14 | 1828 | ||
| XM_006714925.2 | 12 | 1574 | XP_006714988.1 | |
| XM_011534669.2 | 13 | 1730 | XP_011532971.1 | |
| XM_011534670.2 | 12 | 2063 | XP_011532972.1 | |
| XM_011534671.2 | 13 | 1718 | XP_011532973.1 | |
| XM_011534672.2 | 13 | 1516 | XP_011532974.1 | |
| XM_011534674.2 | 10 | 1385 | XP_011532976.1 | |
| XM_011534676.2 | 9 | 1670 | XP_011532978.1 | |
| XM_017009996.1 | 12 | 1599 | XP_016865485.1 | |
| XM_017009997.1 | 12 | 1569 | XP_016865486.1 | |
| XM_017009998.1 | 12 | 2032 | XP_016865487.1 | |
| XM_017009999.1 | 9 | 3475 | XP_016865488.1 | |
| AA862441.1 | ||||
| AF318375.1 | 6 | 2675 | AAL55882.1 | |
| AK023470.1 | 10 | 1690 | ||
| AK292061.1 | 13 | 1700 | ||
| BC008009.1 | 10 | 1292 | AAH08009.1 | |
| BC037567.1 | 13 | 1700 | AAH37567.1 | |
| BC110335.1 | 13 | 1710 | AAI10336.1 | |
| HY267487.1 | 1 | 363 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019404 | Chr.5:178080079 - 178432088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map