Assay Details
Target Gene Details
Entrez Gene ID: | 81607 |
Gene Name: | nectin cell adhesion molecule 4 |
Gene Aliases: |
EDSS1, LNIR, PRR4, PVRL4, nectin-4 |
Location: |
Chr.1:161070991-161089595 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECTIN4 | NM_030916.2 | NP_112178.2 | ||
| XM_005245508.3 | XP_005245565.1 | |||
| XM_011510021.2 | XP_011508323.1 | |||
| XM_011510022.2 | XP_011508324.1 | |||
| XM_011510023.1 | XP_011508325.1 | |||
| AB105191.1 | ||||
| AB755430.1 | ||||
| AF160477.1 | AAF82399.1 | |||
| AF426163.1 | AAL23958.1 | |||
| AK027753.1 | BAB55344.1 | |||
| BC010423.1 | AAH10423.1 | |||
| DB223835.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161729 | Chr.1:161075405 - 161104302 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map