Assay Details
Target Gene Details
Entrez Gene ID: | 3073 |
Gene Name: | hexosaminidase subunit alpha |
Gene Aliases: |
TSD |
Location: |
Chr.15:72343435-72376473 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEXA | NM_000520.5 | 14 | 2119 | NP_000511.2 |
| NM_001318825.1 | 14 | 2152 | NP_001305754.1 | |
| AK222502.1 | 14 | 1632 | BAD96222.1 | |
| AK301000.1 | 14 | 1672 | ||
| BC001138.2 | 12 | 1259 | AAH01138.2 | |
| BC018927.2 | 14 | 1629 | AAH18927.1 | |
| BC021030.1 | 8 | 958 | AAH21030.1 | |
| BC084537.1 | 14 | 1625 | AAH84537.1 | |
| BM668409.1 | ||||
| CR627386.1 | 14 | 2119 | CAH10482.1 | |
| KU178109.1 | ||||
| KU178110.1 | ||||
| KU178111.1 | ||||
| KU178112.1 | ||||
| KU178113.1 | ||||
| M13520.1 | 14 | 1502 | AAA51827.1 |
More Information
Set Membership: |
|

Genomic Map