Assay Details
Target Gene Details
Entrez Gene ID: | 10369 |
Gene Name: | calcium voltage-gated channel auxiliary subunit gamma 2 |
Gene Aliases: |
MRD10 |
Location: |
Chr.22:36560869-36702645 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNG2 | NM_006078.3 | 4 | 681 | NP_006069.1 |
| XM_017028531.1 | 3 | 478 | XP_016884020.1 | |
| AF096322.1 | 4 | 612 | AAD22738.1 | |
| BC069612.1 | 4 | 681 | AAH69612.1 | |
| BC112297.1 | 4 | 672 | AAI12298.1 | |
| BC112299.1 | 4 | 672 | AAI12300.1 | |
| BI919073.1 | 1 | 32 | ||
| CR456414.1 | 4 | 647 | CAG30300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758836 | Chr.22:36510516 - 36645987 on Build GRCh38 | Loss |
|
| esv2760722 | Chr.22:36390921 - 36617980 on Build GRCh38 | Gain |
|
| nsv520497 | Chr.22:36299896 - 36570195 on Build GRCh38 | Gain |
|
| nsv1060004 | Chr.22:36448479 - 36615303 on Build GRCh38 | Gain |
|
| nsv9900 | Chr.22:36562569 - 36565188 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map