Assay Details
Target Gene Details
Entrez Gene ID: | 348995 |
Gene Name: | nucleoporin 43 |
Gene Aliases: |
bA350J20.1, p42 |
Location: |
Chr.6:149724315-149746572 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP43 | NM_198887.2 | NP_942590.1 | ||
| NR_104456.1 | ||||
| XM_005266960.4 | XP_005267017.1 | |||
| XM_005266961.3 | XP_005267018.1 | |||
| XM_005266962.3 | XP_005267019.1 | |||
| XM_011535798.2 | XP_011534100.1 | |||
| XM_011535799.2 | XP_011534101.1 | |||
| AF514997.1 | AAM76708.1 | |||
| AK023349.1 | BAB14536.1 | |||
| AK095994.1 | ||||
| AK301027.1 | ||||
| AK304244.1 | ||||
| AK304505.1 | ||||
| BC040121.1 | ||||
| BC065028.1 | AAH65028.1 | |||
| BG567254.1 | ||||
| BX647097.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2732861 | Chr.6:149681836 - 149764933 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map