Assay Details
Target Gene Details
Entrez Gene ID: | 55691 |
Gene Name: | FERM domain containing 4A |
Gene Aliases: |
CCAFCA, FRMD4, bA295P9.4 |
Location: |
Chr.10:13643706-14330952 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRMD4A | NM_001318336.1 | 24 | 5832 | NP_001305265.1 |
| NM_001318337.1 | 24 | 6072 | NP_001305266.1 | |
| NM_001318338.1 | 14 | 4866 | NP_001305267.1 | |
| NM_018027.4 | 25 | 5772 | NP_060497.3 | |
| AB037715.1 | 25 | 5772 | ||
| BC151244.1 | 25 | 5783 |
Target Gene Details
Entrez Gene ID: | 8559 |
Gene Name: | pre-mRNA processing factor 18 |
Gene Aliases: |
PRP18, hPrp18 |
Location: |
Chr.10:13586811-13645279 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831789 | Chr.10:13536081 - 13716895 on Build GRCh38 | Gain+Loss |
|
| esv3891761 | Chr.10:13544627 - 13695972 on Build GRCh38 | Gain |
|
| esv2761575 | Chr.10:13581531 - 13659502 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map