Assay Details
Target Gene Details
Entrez Gene ID: | 832 |
Gene Name: | capping actin protein of muscle Z-line beta subunit |
Gene Aliases: |
CAPB, CAPPB, CAPZ |
Location: |
Chr.1:19338773-19485641 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPZB | NM_001206540.2 | 10 | 1564 | NP_001193469.1 |
| NM_001206541.2 | 9 | 1871 | NP_001193470.1 | |
| NM_001282162.1 | 9 | 1763 | NP_001269091.1 | |
| NM_004930.4 | 9 | 1451 | NP_004921.1 | |
| AK022336.1 | 1 | 1440 | ||
| AK125085.1 | 9 | 1743 | ||
| AK126650.1 | 1 | 4984 | ||
| BC008095.1 | 3 | 674 | ||
| BQ009850.1 | 1 | 340 | ||
| BQ773230.1 | 1 | 347 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv21e212 | Chr.1:19324106 - 19361938 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map