Assay Details
Target Gene Details
Entrez Gene ID: | 81025 |
Gene Name: | gap junction protein alpha 9 |
Gene Aliases: |
CX58, CX59, GJA10 |
Location: |
Chr.1:38874067-38881626 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJA9 | NM_030772.4 | 1 | 108 | NP_110399.2 |
| AK312811.1 | 1 | 69 | ||
| AL705384.1 | 1 | 108 | ||
| BI463634.1 | 1 | 105 |
Target Gene Details
Entrez Gene ID: | 100527950 |
Gene Name: | GJA9-MYCBP readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:38862490-38881626 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJA9-MYCBP | NR_037633.1 | 1 | 108 | |
| NR_037634.1 | 1 | 108 | ||
| NR_037635.1 | 1 | 108 | ||
| NR_037636.1 | 1 | 108 | ||
| NR_037637.1 | 1 | 108 | ||
| AK123051.1 | 1 | 102 | ||
| FY210901.1 | 1 | 84 | ||
| FY211253.1 | 1 | 84 | ||
| FY211295.1 | 1 | 84 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585774 | Chr.1:38880108 - 38891226 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map