Assay Details
Target Gene Details
Entrez Gene ID: | 10745 |
Gene Name: | putative homeodomain transcription factor 1 |
Gene Aliases: |
PHTF |
Location: |
Chr.1:113697202-113759888 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHTF1 | NM_001323041.1 | 8 | 1398 | NP_001309970.1 |
| NM_001323042.1 | 8 | 882 | NP_001309971.1 | |
| NM_001323043.1 | 8 | 1283 | NP_001309972.1 | |
| NM_001323044.1 | 8 | 1656 | NP_001309973.1 | |
| NM_001323045.1 | 9 | 1367 | NP_001309974.1 | |
| NM_001323046.1 | NP_001309975.1 | |||
| NM_001323047.1 | 8 | 1398 | NP_001309976.1 | |
| NM_001323048.1 | 8 | 1283 | NP_001309977.1 | |
| NM_001323049.1 | 7 | 1572 | NP_001309978.1 | |
| NM_001323050.1 | 9 | 1367 | NP_001309979.1 | |
| NM_001323051.1 | 8 | 1656 | NP_001309980.1 | |
| NM_001323052.1 | 8 | 1283 | NP_001309981.1 | |
| NM_001323053.1 | 8 | 1398 | NP_001309982.1 | |
| NM_006608.2 | 7 | 1187 | NP_006599.2 | |
| NR_136564.1 | 9 | 1482 | ||
| NR_136565.1 | ||||
| NR_136566.1 | 8 | 1398 | ||
| NR_136567.1 | 8 | 1283 | ||
| NR_136568.1 | ||||
| AJ011863.1 | 7 | 766 | CAB51572.1 | |
| AK126773.1 | 1 | 1886 | ||
| AK294749.1 | ||||
| BC000842.1 | AAH00842.1 | |||
| BC002447.2 | 8 | 1331 | AAH02447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587245 | Chr.1:113699035 - 113723249 on Build GRCh38 | Loss |
|
| esv3587240 | Chr.1:113533246 - 114158910 on Build GRCh38 | Loss |
|
| esv3587239 | Chr.1:113510852 - 113866662 on Build GRCh38 | Loss |
|
| nsv831093 | Chr.1:113711545 - 113835613 on Build GRCh38 | Loss |
|
| esv23869 | Chr.1:113320330 - 114358495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map