Assay Details
Target Gene Details
Entrez Gene ID: | 127495 |
Gene Name: | leucine rich repeat containing 39 |
Gene Aliases: |
- |
Location: |
Chr.1:100148448-100178273 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC39 | NM_001256385.1 | 5 | 534 | NP_001243314.1 |
| NM_001256386.1 | 5 | 534 | NP_001243315.1 | |
| NM_001256387.1 | 4 | 477 | NP_001243316.1 | |
| NM_144620.3 | 5 | 517 | NP_653221.1 | |
| AK096892.1 | 5 | 460 | ||
| AL833417.1 | 5 | 482 | ||
| AW961910.1 | 5 | 507 | ||
| AY358131.1 | 4 | 450 | AAQ88498.1 | |
| BC009613.1 | 5 | 459 | AAH09613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map