Assay Details
Target Gene Details
Entrez Gene ID: | 2060 |
Gene Name: | epidermal growth factor receptor pathway substrate 15 |
Gene Aliases: |
AF-1P, AF1P, MLLT5 |
Location: |
Chr.1:51354263-51519354 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPS15 | NM_001159969.1 | 13 | 3984 | NP_001153441.1 |
| NM_001981.2 | 25 | 4853 | NP_001972.1 | |
| XM_005270618.2 | 24 | 4794 | XP_005270675.1 | |
| XM_017000615.1 | 25 | 4995 | XP_016856104.1 | |
| XM_017000616.1 | 24 | 4791 | XP_016856105.1 | |
| XM_017000617.1 | 24 | 4680 | XP_016856106.1 | |
| XM_017000618.1 | 23 | 4590 | XP_016856107.1 | |
| XM_017000619.1 | 26 | 4877 | XP_016856108.1 | |
| XM_017000620.1 | 25 | 5044 | XP_016856109.1 | |
| AA789139.1 | ||||
| BM681433.1 | 1 | 385 |
More Information
Set Membership: |
|

Genomic Map