Assay Details
Target Gene Details
Entrez Gene ID: | 55512 |
Gene Name: | sphingomyelin phosphodiesterase 3 |
Gene Aliases: |
NSMASE2 |
Location: |
Chr.16:68358327-68448703 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPD3 | NM_018667.3 | 9 | 4592 | NP_061137.1 |
| XM_005256031.3 | 9 | 4685 | XP_005256088.1 | |
| XM_005256032.3 | 9 | 4704 | XP_005256089.1 | |
| XM_011523207.1 | 10 | 4685 | XP_011521509.1 | |
| XM_011523208.2 | 8 | 4786 | XP_011521510.1 | |
| XM_011523209.2 | 8 | 4569 | XP_011521511.1 | |
| XM_017023405.1 | 9 | 4511 | XP_016878894.1 | |
| XM_017023406.1 | 10 | 4992 | XP_016878895.1 | |
| XM_017023407.1 | 10 | 4714 | XP_016878896.1 | |
| XM_017023408.1 | 10 | 4668 | XP_016878897.1 | |
| AK026246.1 | 1 | 750 | BAB15409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582280 | Chr.16:68348088 - 68360930 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map