Assay Details
Target Gene Details
Entrez Gene ID: | 22861 |
Gene Name: | NLR family pyrin domain containing 1 |
Gene Aliases: |
CARD7, CIDED, CLR17.1, DEFCAP, DEFCAP-L/S, NAC, NALP1, PP1044, SLEV1, VAMAS1 |
Location: |
Chr.17:5501399-5584512 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP1 | NM_001033053.2 | NP_001028225.1 | ||
| NM_014922.4 | NP_055737.1 | |||
| NM_033004.3 | NP_127497.1 | |||
| NM_033006.3 | NP_127499.1 | |||
| NM_033007.3 | NP_127500.1 | |||
| AB023143.1 | ||||
| AF229059.1 | AAK00748.1 | |||
| AF229060.1 | AAK00749.1 | |||
| AF229061.1 | AAK00750.1 | |||
| AF229062.1 | AAK00751.1 | |||
| AF298548.1 | AAG15254.1 | |||
| AF310105.1 | AAG30288.1 | |||
| AK026393.1 | BAB15469.1 | |||
| AK026398.1 | BAB15470.1 | |||
| AL117470.1 | CAB55945.1 | |||
| AL832400.1 | 1 | 702 |
More Information
Set Membership: |
|

Genomic Map