Assay Details
Target Gene Details
Entrez Gene ID: | 1827 |
Gene Name: | regulator of calcineurin 1 |
Gene Aliases: |
ADAPT78, CSP1, DSC1, DSCR1, MCIP1, RCN1 |
Location: |
Chr.21:34516442-34615142 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCAN1 | NM_001285389.2 | 4 | 1690 | NP_001272318.1 |
| NM_001285391.2 | 4 | 2013 | NP_001272320.2 | |
| NM_001285392.2 | 4 | 1807 | NP_001272321.1 | |
| NM_001285393.2 | 4 | 1744 | NP_001272322.1 | |
| NM_004414.6 | 4 | 2016 | NP_004405.3 | |
| NM_203417.2 | 4 | 1802 | NP_981962.1 | |
| NM_203418.2 | 4 | 1981 | NP_981963.1 | |
| XM_017028283.1 | 5 | 2342 | XP_016883772.1 | |
| AK055845.1 | 3 | 2213 | ||
| AK092184.1 | 4 | 1937 | ||
| AK131569.1 | 4 | 1934 | BAD18702.1 | |
| AY325903.1 | 4 | 2008 | AAP96743.1 | |
| BC002864.2 | 4 | 1776 | AAH02864.1 | |
| U28833.2 | 4 | 1690 | AAB81557.1 | |
| U85265.2 | 4 | 1801 | AAB84370.1 | |
| U85266.2 | 4 | 1744 | AAB84371.2 | |
| U85267.2 | 4 | 1863 | AAB84372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587428 | Chr.21:34515061 - 34543424 on Build GRCh38 | Loss |
|
| dgv7824n54 | Chr.21:34352440 - 34535115 on Build GRCh38 | Gain |
|
| dgv4426n100 | Chr.21:34339322 - 34534919 on Build GRCh38 | Gain |
|
| nsv1055429 | Chr.21:34299853 - 34516938 on Build GRCh38 | Gain |
|
| esv3575386 | Chr.21:34353927 - 34534570 on Build GRCh38 | Gain |
|
| esv2762109 | Chr.21:34350696 - 34531755 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map