Assay Details
Target Gene Details
Entrez Gene ID: | 63874 |
Gene Name: | abhydrolase domain containing 4 |
Gene Aliases: |
ABH4 |
Location: |
Chr.14:22598238-22612864 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD4 | NM_022060.2 | 3 | 358 | NP_071343.2 |
| XM_005267986.4 | 3 | 411 | XP_005268043.1 | |
| AK022878.1 | 3 | 360 | BAB14289.1 | |
| AK293198.1 | 3 | 292 | ||
| AK298127.1 | 3 | 302 | ||
| AK301315.1 | 3 | 296 | ||
| BC024779.1 | 3 | 306 | AAH24779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832746 | Chr.14:22506563 - 22681586 on Build GRCh38 | Gain |
|
| nsv563998 | Chr.14:22414183 - 22879743 on Build GRCh38 | Gain |
|
| nsv832748 | Chr.14:22515242 - 22630030 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map