Assay Details
Target Gene Details
Entrez Gene ID: | 1393 |
Gene Name: | corticotropin releasing hormone binding protein |
Gene Aliases: |
CRF-BP, CRFBP |
Location: |
Chr.5:76952713-76981143 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRHBP | NM_001882.3 | NP_001873.2 | ||
| CB959505.1 |
Target Gene Details
Entrez Gene ID: | 170591 |
Gene Name: | S100 calcium binding protein Z |
Gene Aliases: |
Gm625, S100-zeta |
Location: |
Chr.5:76849953-76952882 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100Z | XM_011543241.2 | XP_011541543.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830355 | Chr.5:76898897 - 77072195 on Build GRCh38 | Gain |
|
| nsv1027236 | Chr.5:76895111 - 77118908 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map