Assay Details
Target Gene Details
Entrez Gene ID: | 718 |
Gene Name: | complement component 3 |
Gene Aliases: |
AHUS5, ARMD9, ASP, C3a, C3b, CPAMD1, HEL-S-62p |
Location: |
Chr.19:6677835-6720682 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 34 - Intron 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3 | NM_000064.3 | NP_000055.2 | ||
| AK094728.1 | ||||
| AK304071.1 | ||||
| BC150179.1 | ||||
| BC150200.1 | ||||
| EU794602.1 | ||||
| K02765.1 | AAA85332.1 |
More Information
Set Membership: |
|

Genomic Map