Assay Details
Target Gene Details
Entrez Gene ID: | 85300 |
Gene Name: | ATCAY, caytaxin |
Gene Aliases: |
BNIP-H, CLAC |
Location: |
Chr.19:3880620-3928082 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATCAY | NM_033064.4 | NP_149053.1 | ||
| AB058775.1 | ||||
| AK027889.1 | ||||
| AK092309.1 | BAC03859.1 | |||
| AK125457.1 | ||||
| AK299522.1 | ||||
| AY220297.1 | AAO63019.1 | |||
| BC026217.1 | AAH26217.1 | |||
| BX505461.1 | ||||
| DA151785.1 | ||||
| DA778097.1 | ||||
| KU178814.1 | ||||
| KU178815.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833715 | Chr.19:3786758 - 3977144 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map