Assay Details
Target Gene Details
Entrez Gene ID: | 81566 |
Gene Name: | cysteine and serine rich nuclear protein 2 |
Gene Aliases: |
C12orf2, C12orf22, FAM130A1, PPP1R72, TAIP-12 |
Location: |
Chr.12:51061205-51083671 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSRNP2 | NM_030809.2 | 5 | 2236 | NP_110436.1 |
| NR_045072.1 | 5 | 2329 | ||
| XM_006719621.2 | 6 | 2044 | XP_006719684.1 | |
| XM_017019991.1 | 2 | 1448 | XP_016875480.1 | |
| AJ298133.1 | 5 | 2115 | CAC22252.1 | |
| BC017221.2 | 5 | 1921 | AAH17221.1 |
Target Gene Details
Entrez Gene ID: | 25875 |
Gene Name: | LETM1 domain containing 1 |
Gene Aliases: |
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2 |
Location: |
Chr.12:51048230-51068984 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LETMD1 | XM_011538163.2 | XP_011536465.1 | ||
| XM_017019168.1 | XP_016874657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629498 | Chr.12:51040936 - 51066446 on Build GRCh38 | Gain |
|
| esv34039 | Chr.12:50932919 - 51278127 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map