Assay Details
Target Gene Details
Entrez Gene ID: | 54934 |
Gene Name: | KAT8 regulatory NSL complex subunit 2 |
Gene Aliases: |
C12orf41, NSL2 |
Location: |
Chr.12:48653211-48682252 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANSL2 | NM_017822.3 | 3 | 396 | NP_060292.3 |
| AK000443.1 | 3 | 355 | BAA91169.1 | |
| AK022732.1 | 2 | 134 | BAB14211.1 | |
| AK094528.1 | 2 | 919 | ||
| BC009746.2 | 3 | 364 | AAH09746.1 | |
| BC013900.2 | 2 | 316 | AAH13900.1 | |
| DA278834.1 | 3 | 398 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519053 | Chr.12:48671944 - 48685687 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map