Assay Details
Target Gene Details
Entrez Gene ID: | 100505854 |
Gene Name: | Alu-mediated CDKN1A/p21 transcriptional regulator (non-protein coding) |
Gene Aliases: |
RSBN1L-AS1 |
Location: |
Chr.7:77657660-77697345 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APTR | NR_038361.1 | 1 | 464 | |
AF290475.1 | 1 | 471 |
Target Gene Details
Entrez Gene ID: | 222194 |
Gene Name: | round spermatid basic protein 1 like |
Gene Aliases: |
- |
Location: |
Chr.7:77696426-77779803 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RSBN1L | NM_198467.2 | 1 | 457 | NP_940869.2 |
BC046353.1 | 1 | 415 | AAH46353.1 | |
BC059402.1 | 1 | 440 | AAH59402.1 | |
CN311934.1 | 1 | 462 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831039 | Chr.7:77564611 - 77722887 on Build GRCh38 | Gain |
![]() ![]() ![]() |
