Assay Details
Target Gene Details
Entrez Gene ID: | 8848 |
Gene Name: | TSC22 domain family member 1 |
Gene Aliases: |
Ptg-2, TGFB1I4, TSC22 |
Location: |
Chr.13:44432143-44576565 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D1 | NM_001243799.1 | 1 | 134 | NP_001230728.1 |
| NM_183422.3 | 1 | 134 | NP_904358.2 | |
| XM_005266583.4 | XP_005266640.1 | |||
| XM_017020810.1 | XP_016876299.1 | |||
| XM_017020811.1 | XP_016876300.1 | |||
| AB082525.1 | 1 | 134 |
Target Gene Details
Entrez Gene ID: | 641467 |
Gene Name: | TSC22D1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:44575896-44580432 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D1-AS1 | NR_038381.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs111945998] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map