Assay Details
Target Gene Details
Entrez Gene ID: | 6698 |
Gene Name: | small proline rich protein 1A |
Gene Aliases: |
SPRK |
Location: |
Chr.1:152984088-152985814 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPRR1A | NM_001199828.1 | 2 | 454 | NP_001186757.1 |
| NM_005987.3 | 2 | 440 | NP_005978.2 | |
| AW204307.1 | 1 | 218 | ||
| S73288.1 | 1 | 117 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998873 | Chr.1:152896651 - 153002782 on Build GRCh38 | Gain |
|
| nsv831592 | Chr.1:152842578 - 153024686 on Build GRCh38 | Gain |
|
| nsv547899 | Chr.1:152699529 - 153272606 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map