Assay Details
Target Gene Details
Entrez Gene ID: | 64763 |
Gene Name: | zinc finger protein 574 |
Gene Aliases: |
FP972 |
Location: |
Chr.19:42068477-42081567 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF574 | NM_022752.5 | 2 | 2586 | NP_073589.4 |
| XM_005259159.2 | 1 | 2449 | XP_005259216.1 | |
| XM_011527228.2 | 2 | 2779 | XP_011525530.1 | |
| XM_011527229.2 | 2 | 2502 | XP_011525531.1 | |
| XM_011527230.2 | 2 | 2454 | XP_011525532.1 | |
| XM_017027149.1 | 2 | 2922 | XP_016882638.1 | |
| AF447880.1 | 2 | 2530 | AAQ04655.1 | |
| AK025712.1 | 1 | 1694 | BAB15225.1 | |
| AK027895.1 | 2 | 2489 | ||
| AK074788.1 | 1 | 1147 | BAC11210.1 | |
| AK131369.1 | 2 | 2922 | BAD18520.1 | |
| AK131418.1 | 2 | 3072 | BAD18565.1 | |
| AK225342.1 | 1 | 1694 | ||
| AL137516.1 | 1 | 1663 | CAB70782.1 | |
| BC001184.1 | 2 | 2475 | AAH01184.2 | |
| BC071962.1 | 1 | 1239 | AAH71962.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map