Assay Details
Target Gene Details
Entrez Gene ID: | 6249 |
Gene Name: | CAP-Gly domain containing linker protein 1 |
Gene Aliases: |
CLIP, CLIP-170, CLIP170, CYLN1, RSN |
Location: |
Chr.12:122271432-122422956 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLIP1 | NM_001247997.1 | NP_001234926.1 | ||
| NM_002956.2 | NP_002947.1 | |||
| NM_198240.1 | NP_937883.1 | |||
| XM_005253593.2 | XP_005253650.1 | |||
| XM_006719552.3 | XP_006719615.1 | |||
| XM_017019786.1 | XP_016875275.1 | |||
| XM_017019787.1 | XP_016875276.1 | |||
| XM_017019788.1 | XP_016875277.1 | |||
| XM_017019789.1 | XP_016875278.1 | |||
| XM_017019790.1 | XP_016875279.1 | |||
| XM_017019791.1 | XP_016875280.1 | |||
| XM_017019792.1 | XP_016875281.1 | |||
| AB209115.1 | BAD92352.1 | |||
| AK123769.1 | ||||
| AK126990.1 | ||||
| BC114213.1 | AAI14214.1 | |||
| BC117209.1 | ||||
| BC126305.1 | ||||
| M97501.1 | AAA35693.1 | |||
| X64838.1 | CAA46050.1 |
More Information
Set Membership: |
|

Genomic Map