Assay Details
Target Gene Details
Entrez Gene ID: | 23524 |
Gene Name: | serine/arginine repetitive matrix 2 |
Gene Aliases: |
300-KD, CWF21, Cwc21, HSPC075, SRL300, SRm300 |
Location: |
Chr.16:2752329-2771412 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRM2 | NM_016333.3 | 11 | 4379 | NP_057417.3 |
| XM_005255226.1 | 11 | 4357 | XP_005255283.1 | |
| AB002322.3 | 11 | 4031 | ||
| AB016087.1 | 6 | 3201 | BAA83713.1 | |
| AB016090.1 | 1 | 1209 | BAA83716.1 | |
| AB016092.1 | 11 | 4055 | BAA83718.1 | |
| AF201422.1 | 11 | 4059 | AAF21439.1 | |
| BC041155.1 | AAH41155.1 | |||
| BX647088.1 | 10 | 3925 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss |
|
| nsv471073 | Chr.16:2687070 - 2773569 on Build GRCh38 | Loss |
|
| nsv833123 | Chr.16:2561247 - 2764988 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map