Assay Details
Target Gene Details
Entrez Gene ID: | 389362 |
Gene Name: | proteasome assembly chaperone 4 |
Gene Aliases: |
C6orf86, PAC4, bA506K6.2 |
Location: |
Chr.6:3254325-3308224 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMG4 | XM_011514593.2 | XP_011512895.1 | ||
| AK095683.1 |
Target Gene Details
Entrez Gene ID: | 63027 |
Gene Name: | solute carrier family 22 member 23 |
Gene Aliases: |
C6orf85 |
Location: |
Chr.6:3268973-3456574 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A23 | NM_001286455.1 | 10 | 1925 | NP_001273384.1 |
| NM_015482.1 | 10 | 2384 | NP_056297.1 | |
| NM_021945.6 | 11 | 2170 | NP_068764.3 | |
| NR_104448.1 | ||||
| XM_011514801.2 | 11 | 2471 | XP_011513103.1 | |
| XM_017011180.1 | 11 | 2948 | XP_016866669.1 | |
| XM_017011181.1 | 9 | 1697 | XP_016866670.1 | |
| XM_017011182.1 | 8 | 1979 | XP_016866671.1 | |
| XM_017011184.1 | 8 | 1634 | XP_016866673.1 | |
| XM_017011186.1 | 11 | 1867 | XP_016866675.1 | |
| AK000691.1 | 1 | 218 | ||
| AK172770.1 | 7 | 1480 | BAD18753.1 | |
| BC128580.1 | 10 | 1877 | ||
| BC128581.1 |
More Information
Set Membership: |
|

Genomic Map