Assay Details
Target Gene Details
Entrez Gene ID: | 51661 |
Gene Name: | FK506 binding protein 7 |
Gene Aliases: |
FKBP23, PPIase |
Location: |
Chr.2:178463664-178479878 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP7 | NM_001135212.1 | NP_001128684.1 | ||
| NM_181342.2 | NP_851939.1 | |||
| XM_005246638.4 | XP_005246695.1 | |||
| XM_011511348.2 | XP_011509650.1 | |||
| XM_011511349.2 | XP_011509651.1 | |||
| AF092137.1 | AAD40379.1 | |||
| AF100751.1 | AAD43015.1 | |||
| AK292145.1 | ||||
| AK299215.1 | ||||
| AY353086.1 | AAQ57208.1 | |||
| AY359015.1 | AAQ89374.1 | |||
| BC009711.1 | AAH09711.1 | |||
| BF210667.1 | ||||
| BT007122.1 | AAP35786.1 | |||
| DA020387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834467 | Chr.2:178393413 - 178566238 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map