Assay Details
Target Gene Details
Entrez Gene ID: | 7001 |
Gene Name: | peroxiredoxin 2 |
Gene Aliases: |
HEL-S-2a, NKEF-B, NKEFB, PRP, PRX2, PRXII, PTX1, TDPX1, TPX1, TSA |
Location: |
Chr.19:12796820-12801910 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDX2 | NM_005809.5 | NP_005800.3 | ||
| AK022395.1 | 1 | 1617 | ||
| AK293957.1 | ||||
| BC000452.2 | AAH00452.1 | |||
| BC003022.1 | AAH03022.1 | |||
| BC039428.1 | AAH39428.1 | |||
| BG705975.1 | ||||
| BP248561.1 | ||||
| CR450356.1 | CAG29352.1 | |||
| CR541789.1 | CAG46588.1 | |||
| EU668325.1 | ||||
| L19185.1 | AAA50465.1 | |||
| X82321.1 | CAA57764.1 | |||
| Z22548.1 | CAA80269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv1160586 | Chr.19:12597480 - 12823909 on Build GRCh38 | Duplication |
|
| dgv138n111 | Chr.19:12627853 - 12810372 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map