Assay Details
Target Gene Details
Entrez Gene ID: | 54107 |
Gene Name: | DNA polymerase epsilon 3, accessory subunit |
Gene Aliases: |
CHARAC17, CHRAC17, YBL1, p17 |
Location: |
Chr.9:113407235-113410749 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLE3 | NM_001278255.1 | 4 | 429 | NP_001265184.1 |
| NM_017443.4 | 4 | 459 | NP_059139.3 | |
| NR_027261.1 | 3 | 593 | ||
| AF226077.1 | 3 | 211 | AAF72417.1 | |
| AK074629.1 | 3 | 303 | BAC11099.1 | |
| AK074762.1 | 4 | 382 | BAC11190.1 | |
| AK074782.1 | 3 | 292 | BAC11206.1 | |
| AK092840.1 | 2 | 587 | ||
| AK096050.1 | 3 | 295 | ||
| AY720898.1 | 3 | 211 | AAU15052.1 | |
| BC003166.1 | 3 | 282 | AAH03166.1 | |
| BC004170.2 | 3 | 282 | AAH04170.1 | |
| DA477404.1 | 4 | 355 | ||
| DA521953.1 | ||||
| DA908447.1 | 4 | 460 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621540 | Chr.9:113279513 - 113421563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map