Assay Details
Target Gene Details
Entrez Gene ID: | 78992 |
Gene Name: | Yip1 domain family member 2 |
Gene Aliases: |
FinGER2 |
Location: |
Chr.19:10922185-10929012 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIPF2 | NM_001321439.1 | NP_001308368.1 | ||
| NM_001321440.1 | NP_001308369.1 | |||
| NM_024029.4 | NP_076934.1 | |||
| XM_011528270.2 | XP_011526572.1 | |||
| XM_011528271.2 | XP_011526573.1 | |||
| BC000056.2 | AAH00056.1 | |||
| BC013014.2 | AAH13014.1 | |||
| BM554546.1 | ||||
| HY020827.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833749 | Chr.19:10881255 - 11027578 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map