Assay Details
Target Gene Details
Entrez Gene ID: | 7150 |
Gene Name: | topoisomerase (DNA) I |
Gene Aliases: |
TOPI |
Location: |
Chr.20:41028822-41124487 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOP1 | NM_003286.2 | 1 | 90 | NP_003277.1 |
| AK225095.1 | 1 | 89 | ||
| AK292943.1 | 1 | 89 | ||
| AK310508.1 | 1 | 69 | ||
| AK310516.1 | 1 | 90 | ||
| BC004475.2 | 1 | 72 | AAH04475.1 | |
| BC067349.1 | 1 | 69 | AAH67349.1 | |
| J03250.1 | 1 | 55 | AAA61207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064246 | Chr.20:40261343 - 42254320 on Build GRCh38 | Loss |
|
| nsv833982 | Chr.20:40986487 - 41110831 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map