Assay Details
Target Gene Details
Entrez Gene ID: | 79901 |
Gene Name: | cytochrome b reductase 1 |
Gene Aliases: |
CYB561A2, DCYTB, FRRS3 |
Location: |
Chr.2:171522247-171558133 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYBRD1 | NM_001127383.1 | NP_001120855.1 | ||
| NM_001256909.1 | NP_001243838.1 | |||
| NM_024843.3 | NP_079119.3 | |||
| AK027115.1 | BAB15661.1 | |||
| AK128495.1 | BAC87466.1 | |||
| AK225459.1 | ||||
| AK291731.1 | ||||
| AK301485.1 | ||||
| AK316588.1 | ||||
| AL136693.1 | CAB66628.1 | |||
| BC065290.1 | AAH65290.1 | |||
| CB266594.1 | ||||
| CX866825.1 | ||||
| DB235339.1 |
More Information
Set Membership: |
|

Genomic Map