Assay Details
Target Gene Details
Entrez Gene ID: | 57453 |
Gene Name: | DS cell adhesion molecule like 1 |
Gene Aliases: |
DSCAM2 |
Location: |
Chr.11:117427772-117797261 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSCAML1 | NM_020693.3 | 25 | 4526 | NP_065744.2 |
| XM_011542917.1 | 25 | 4556 | XP_011541219.1 | |
| XM_011542918.1 | 26 | 4244 | XP_011541220.1 | |
| XM_011542919.2 | 20 | 3199 | XP_011541221.1 | |
| XM_011542920.2 | 20 | 3246 | XP_011541222.1 | |
| XM_011542921.2 | 16 | 2803 | XP_011541223.1 | |
| XM_011542924.2 | 15 | 2721 | XP_011541226.1 | |
| XM_011542925.2 | 15 | 3395 | XP_011541227.1 | |
| AB032958.2 | 25 | 4461 | BAA86446.2 | |
| AF304304.1 | 25 | 4356 | AAN32613.1 | |
| AF304305.1 | 23 | 3723 | AAN32614.1 | |
| AF334384.1 | 25 | 4438 | AAL57166.1 | |
| AF491813.1 | 25 | 4526 | AAM09558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052364 | Chr.11:117390271 - 117479621 on Build GRCh38 | Gain |
|
| nsv556458 | Chr.11:117425576 - 117471747 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map