Assay Details
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPHN | XM_017020925.1 | XP_016876414.1 |
Target Gene Details
Entrez Gene ID: | 64398 |
Gene Name: | membrane palmitoylated protein 5 |
Gene Aliases: |
PALS1 |
Location: |
Chr.14:67241295-67336061 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP5 | NM_001256550.1 | 1 | 164 | NP_001243479.1 |
| NM_022474.3 | 1 | 164 | NP_071919.2 | |
| XM_011537086.2 | 1 | 64 | XP_011535388.1 | |
| AI245706.1 | 1 | 164 | ||
| AK057160.1 | ||||
| AK098373.1 | 1 | 128 | BAC05295.1 | |
| AK291240.1 | 1 | 145 | ||
| AL832326.1 | 1 | 61 | CAD38620.1 | |
| AL832578.2 | CAD89937.1 | |||
| BC053366.1 | 1 | 99 | AAH53366.1 | |
| BC095485.1 | 1 | 350 | AAH95485.1 | |
| BC129933.1 | 1 | 49 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581256 | Chr.14:67234632 - 67246628 on Build GRCh38 | Loss |
|
| esv2751285 | Chr.14:66898030 - 67420064 on Build GRCh38 | Gain |
|
| nsv832821 | Chr.14:67123701 - 67361452 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs200541606] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map