Assay Details
Target Gene Details
Entrez Gene ID: | 152185 |
Gene Name: | spindle and centriole associated protein 1 |
Gene Aliases: |
CCDC52, SPICE |
Location: |
Chr.3:113442718-113515187 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPICE1 | NM_144718.3 | NP_653319.1 | ||
| XM_011512461.2 | XP_011510763.1 | |||
| XM_017005775.1 | XP_016861264.1 | |||
| XM_017005776.1 | XP_016861265.1 | |||
| XM_017005777.1 | XP_016861266.1 | |||
| XM_017005778.1 | XP_016861267.1 | |||
| AK129575.1 | ||||
| AL832148.1 | CAD91167.1 | |||
| AL832327.2 | ||||
| AY099107.1 | AAM34495.1 | |||
| BC019232.1 | AAH19232.1 | |||
| BC036951.1 | AAH36951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map