Assay Details
Target Gene Details
Entrez Gene ID: | 54946 |
Gene Name: | solute carrier family 41 member 3 |
Gene Aliases: |
SLC41A1-L2 |
Location: |
Chr.3:126006357-126101550 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC41A3 | NM_001008485.1 | NP_001008485.1 | ||
| NM_001008486.1 | NP_001008486.1 | |||
| NM_001008487.1 | 10 | 1580 | NP_001008487.1 | |
| NM_001164475.1 | 10 | 1282 | NP_001157947.1 | |
| NM_017836.3 | 11 | 1661 | NP_060306.3 | |
| XM_005247559.1 | XP_005247616.1 | |||
| XM_005247562.1 | XP_005247619.1 | |||
| XM_005247563.2 | XP_005247620.1 | |||
| XM_005247564.1 | 11 | 1539 | XP_005247621.1 | |
| XM_005247565.2 | XP_005247622.1 | |||
| XM_006713681.2 | XP_006713744.1 | |||
| XM_011512945.1 | XP_011511247.1 | |||
| XM_017006705.1 | XP_016862194.1 | |||
| XM_017006706.1 | 11 | 1555 | XP_016862195.1 | |
| XM_017006707.1 | 11 | 1578 | XP_016862196.1 | |
| XM_017006708.1 | 10 | 1386 | XP_016862197.1 | |
| XM_017006709.1 | 10 | 1441 | XP_016862198.1 | |
| AK000480.1 | BAA91192.1 | |||
| AK021925.1 | 11 | 1347 | BAB13938.1 | |
| AK022780.1 | BAB14241.1 | |||
| AK091671.1 | 12 | 1465 | BAC03717.1 | |
| AK093380.1 | 10 | 1577 | ||
| AK094051.1 | 6 | 1670 | ||
| AK128054.1 | ||||
| AK298070.1 | 10 | 1291 | ||
| AW938624.1 | ||||
| BC009039.2 | AAH09039.1 | |||
| BC028241.1 | AAH28241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009333 | Chr.3:126006809 - 126041785 on Build GRCh38 | Loss |
|
| nsv591465 | Chr.3:125283331 - 126140524 on Build GRCh38 | Gain |
|
| nsv829716 | Chr.3:125928159 - 126116976 on Build GRCh38 | Gain+Loss |
|
| nsv1011457 | Chr.3:125287676 - 126146923 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map