Assay Details
Target Gene Details
Entrez Gene ID: | 2027 |
Gene Name: | enolase 3 |
Gene Aliases: |
GSD13, MSE |
Location: |
Chr.17:4949182-4957131 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENO3 | NM_001193503.1 | NP_001180432.1 | ||
| NM_001976.4 | NP_001967.3 | |||
| NM_053013.3 | NP_443739.3 | |||
| XM_005256521.2 | XP_005256578.1 | |||
| XM_011523729.1 | XP_011522031.1 | |||
| XM_017024346.1 | XP_016879835.1 | |||
| AK300662.1 | ||||
| AK300673.1 | ||||
| AK300709.1 | ||||
| AK309864.1 | ||||
| AU143784.1 | ||||
| BC017249.2 | AAH17249.1 | |||
| BJ995304.1 | ||||
| DA082822.1 | ||||
| DA566866.1 | ||||
| X16504.1 | CAA34513.1 | |||
| X51957.1 | CAA36216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833350 | Chr.17:4884713 - 5060776 on Build GRCh38 | Loss |
|
| nsv833349 | Chr.17:4853631 - 4991300 on Build GRCh38 | Loss |
|
| nsv833347 | Chr.17:4785297 - 4955981 on Build GRCh38 | Loss |
|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs151153333] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map