Assay Details
Target Gene Details
Entrez Gene ID: | 254268 |
Gene Name: | AKNA domain containing 1 |
Gene Aliases: |
C1orf62 |
Location: |
Chr.1:108815898-108858242 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKNAD1 | NM_152763.4 | 6 | 1611 | NP_689976.2 |
| NR_049760.1 | 7 | 1752 | ||
| AK095517.1 | 7 | 1738 | BAC04565.1 | |
| AL832216.1 | 6 | 1601 | ||
| BC030647.2 | 6 | 1608 | AAH30647.1 | |
| BC150642.1 | 5 | 1366 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587140 | Chr.1:108842339 - 108891089 on Build GRCh38 | Gain |
|
| nsv517381 | Chr.1:108825322 - 108843844 on Build GRCh38 | Loss |
|
| nsv1013192 | Chr.1:108803093 - 108868275 on Build GRCh38 | Gain |
|
| esv3587139 | Chr.1:108842102 - 108869065 on Build GRCh38 | Gain |
|

Genomic Map