Assay Details
Target Gene Details
Entrez Gene ID: | 84897 |
Gene Name: | transforming growth factor beta regulator 1 |
Gene Aliases: |
NIAM, TB-5 |
Location: |
Chr.11:124622599-124635926 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBRG1 | NM_032811.2 | 9 | 2160 | NP_116200.2 |
| NR_016021.1 | 8 | 2023 | ||
| XM_005271709.3 | 6 | 1966 | XP_005271766.1 | |
| XM_017018442.1 | 6 | 2981 | XP_016873931.1 | |
| AK027527.1 | 8 | 1994 | BAB55177.1 | |
| AK027766.1 | 1 | 330 | ||
| AK074140.1 | 8 | 1747 | BAB84966.1 | |
| AL713631.1 | 5 | 1265 | CAD28450.1 | |
| AL831881.1 | 3 | 1000 | CAH10595.1 | |
| BC109269.2 | 7 | 1642 | AAI09270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045156 | Chr.11:124565356 - 124705852 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map